Learn More
Abeba™ Reflexor™ 6912 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 6912/37
Additional Details : Weight : 0.85000kg
 View more versions of this product
Product Code. 15662428
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRC
  • Smooth leather
  • Black
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3565/ 3568)
  • Antistatic
  • Lining microfiber, respiration absorbing
  • 3 adjustable instep straps
  • velcro fastening
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRC
Microfiber
37
Polyurethane
Black
Leather
Slip-resistant
6912