missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Reinforced Polypropylene Mandatory Signs
Marque: Brady™ 252159
Informations supplémentaires : Weight : 0.05300kg
Spécification
Polypropylene | |
Reinforced Polypropylene | |
120°C | |
Round | |
Mandatory Sign |
1 Ea. | |
200 mm | |
0.369 mm | |
Use Overshoes | |
White on Blue |