Learn More
Brady™ Reinforced Polypropylene Warning Labels: Dangerous Electrical Voltage
10.65€ - 24.60€
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10080904
|
Brady™
252214 |
200W x 200 mmH |
10.65€
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
11734844
|
Brady™
252215 |
315W x 315 mmH |
24.60€
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Specifications
Triangular |