Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Brand:  Thermo Scientific™ R30161901
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12729300
  • 100.00€ / Each
Estimated Shipment: 06-07-2021
Add to basket
Description
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Specifications
Positive
Agglutinating Sera
flagellar H antigens of Salmonella strains by slide and/or tube agglutination test
2mL
Salmonella H
Salmonella H (lv)
2mL/Vial