Learn More
Thermo Scientific™ Remel™ BactiDrop India Ink
Remel BactiDrop India Ink is for microscopic examination of encapsulated microorganisms.
Marke:  Thermo Scientific™ R21518
Weitere Details : Weight : 1.02060kg
Artikelnummer. 12729240
  • 138.00€ / Each
Estimated Shipment: 24-06-2024
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Remel BactiDrop India Ink is for microscopic examination of encapsulated microorganisms.
Ordering Alerts: Supplied in crushable ampules enclosed in a protective plastic sheath with a convenient dropper dispenser tip.
Spezifikation
Spezifikation
0.75 mL/Ampule
India Ink
50 Pk.
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote