Learn More
Thermo Scientific™ Remel™ Calibrated Inoculating Loops
Thermo Scientific™ Remel Calibrated Inoculating Loops come in a variety of materials: platinum, twisted nichrome and plastic.Some with color coded with insulated handle.
Brand: Thermo Scientific™ R501511
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ Remel Calibrated Inoculating Loops are available in platinum, twisted nichrome and and plastic. Some are color coded with insulated handle.
- Twisted nichrome or plastic
- With or without insulated handle
- Color-coded
Specifications
Plastic, Nichrome | |
Yellow | |
32/Pk. |
10 μL | |
Inoculating Loop |