Learn More
Labconco™ Replacement Bag for Bag-In/Bag-Out HEPA/ULPA Filter
For use with 4-foot XPert™, RXPert™ and XPert™ Nano enclosures. Labconco™ Replacement Bag for Bag-In/Bag-Out HEPA/ULPA Filter make for easy replacement and disposal of filters without allowing particulates to be dispersed.
Brand:  Labconco™ 3776025
Additional Details : Weight : 4.10000kg
Product Code. 15878946
  • 385.00€ / Each
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
For use with the following types of particulate filters:
  • XPert, RXPert Balance and XPert Bulk Powder Enclosures
  • XPert Nano Enclosures
Specifications
Specifications
Particulate HEPA/ULPA
96 in.
243.8 cm
Filter Bags
Filter Installation, Replacement and Disposal
XPert™, RXPert™ and XPert™ Nano enclosures
96 in. (Circumference)
243.8 cm (Circumference)
48 in.