Learn More
Cole-Parmer™ Replacement Ionic Strength adjustors
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors
Brand: Cole-Parmer™ 27503-51
Additional Details : Weight : 0.01000kg
Beschreibung
- Replacement ionic strength adjustor
- 5 M NaNO3
- Replacement ionic strength adjustors come in 475mL bottles (except model 27503-58, which comes in a one-gallon bottle)
Spezifikation
Reference Electrode: 5 M NaNO3 | |
Replacement Ionic Strength Adjustors |
Sicherheit und Handhabung
- SODIUM NITRATE ADJUSTOR 5M
Signalwort
- Warning
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
Gefahrenhinweis
- H319-Causes serious eye irritation.
Sicherheitshinweis
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Ergänzende Informationen
- MIXTURE LIST-Contains : sodium nitrate