Learn More
Hamilton™ Replacement Needles
Click to view available options
Diameter (Metric) Outer:
0.17 mm
0.23 mm
Unit Size:
Pack of 3
Specifications
Specifications
| Diameter (Metric) Outer | 0.17 mm |
| Product Type | Replacement Needle |
| For Use With (Equipment) | Grab N-Column |