Learn More
Trajan Scientific and Medical™ Replacement Needles
Product Code. 12732237
Click to view available options
Unit Size:
Pack of 5
Product Code. 12732237
Brand: Trajan Scientific and Medical™ 037160
For use with 10μL Syringes
  • For use with 10μL syringes
  • Supplied in packs of 5
Specifications
Diameter (Metric) Outer 0.47 mm
Product Type Replacement Needle
Length (Metric) Needle 115 mm
For Use With (Equipment) Syringe (10μL)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.