Learn More
Trajan Scientific and Medical™ Replacement Plungers for Agilent Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031823
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15278543
  • 71.40€ / Each
Estimated Shipment: 20-06-2024
to see stock.
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
23 to 26 G
0.63/0.47 mm
100 μL
0.24 mm
Fixed Needle
42 mm