Learn More
Trajan Scientific and Medical™ Replacement Plungers for Agilent Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031808
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218533
  • 72.60€ / Pack of 2
Estimated Shipment: 14-06-2024
to see stock.
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Replacement plunger
0.63/0.47 mm
Fixed Needle
Cone
23 G
0.63 mm
10 μL
Gas Tight Syringe