Learn More
Trajan Scientific and Medical™ Replacement Plungers for Agilent Syringes
Replacement plungers for gas tight syringes.
Brand: Trajan Scientific and Medical™ 031823
Additional Details : Weight : 0.01000kg
Description
Designed to match the specific syringe barrel to maintain a gas tight sealSpecifications
23 to 26 G | |
0.63/0.47 mm | |
100 μL |
0.24 mm | |
Fixed Needle | |
42 mm |