Learn More
Trajan Scientific and Medical™ Replacement Probe for Headspace/Soil Gas Syringes
Click to view available options
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Headspace/soil gas analysis syringes
Specifications
Specifications
| Description | Replacement Probe |
| Diameter (Metric) Inner Needle | 0.75 mm |
| Diameter (Metric) Outer Needle | 1.587 mm |
| Length (Metric) | 60 mm |
| Volume (Metric) | 5 to 10 mL |
| For Use With (Equipment) | Headspace/Soil Gas Syringes |