Learn More
Trajan™ SilTite™ FingerTite Ferrule
Click to view available options
Diameter (Metric) Inside:
0.4 mm
0.5 mm
0.7 mm
Unit Size:
Pack of 10
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Specifications
| Type | GC Ferrule |
| Quantity | 10/Pk. |
| Diameter (Metric) Inside | 0.7 mm |