Learn More
Sartorius™ ReproEasy Performance Test Pads
Test pads for the periodic review of the performance of MA37/160 moisture analyzer.
67.20€
Specifications
For Use With (Equipment) | MA37/160 Moisture Analyzer |
---|---|
Type | Moisture Analyzer Test Pads, 10/Pk. |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15689963
|
Sartorius™
YHP01MA |
Moisture Analyzer Test Pads, 10/Pk. |
67.20€
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Test pads for the periodic review of the performance of MA37/160 moisture analyzer.
- Allows you to quickly confirm whether a device is working properly and delivers correct results.
- Use together with the performance test function in moisture analyzer.
Specifications
MA37/160 Moisture Analyzer | |
Moisture Analyzer Test Pads, 10/Pk. |