Learn More
Invitrogen™ RET [G691S] Recombinant Human Protein
Human RET G691S, C-terminal fragment, amino acids H658-S1114 (as in NCBI/Protein entry NP_066124.1) with a G691S mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
Brand: Invitrogen™ PV6215
Additional Details : Weight : 0.01000kg
Description
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
80.5kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 658-1114 | |
Recombinant |
50mM HEPES pH 7.5, 100mM NaCl, 5mM DTT, 15mM reduced glutathione, 20% glycerol | |
100μg | |
CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET51, RET-ELE1 | |
Human | |
GST Tag, His Tag |