missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RET [Y791F] Recombinant Human Protein

The receptor tyrosine kinase RET is a member of the cadherin superfamily.

Brand:  Invitrogen™ PR8194A

Additional Details : Weight : 0.01000kg

Product Code. 15428663

  • 2915.00€ / 100µg
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RET plays a crucial role in neural crest development, and can undergo oncogenic activation by cytogenetic rearrangement. The Y791F mutant has been observed clinical in association with cancer.

Specifications

Specifications

NP_055124, Y791F
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
RET Y791F
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
79kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
RET Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions