Learn More
Bochem™ Retort Clamps
Bochem™ Retort Clamps with adjustable span width.
32.35€ - 89.50€
Specifications
Diameter (Metric) | 8 mm |
---|---|
Length (Metric) | 125 mm |
Size (Metric) Grip | 8 to 30 mm |
Product Code | Brand | Material | Jaw Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Jaw Type | Price | Quantity & Availability | ||||
10712523
|
Bochem™
B08307036 |
Stainless Steel (18/10) | Transparent Silicone-coated |
89.50€
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
10491411
|
Bochem™
B08235037 |
Laboral | Red PVC-coated |
32.35€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
||||
Description
- Stainless Steel
- Adjustable
Specifications
8 mm | |
125 mm | |
8 to 30 mm |