Learn More
RETSCH MM 400 Mixer Mills: Grinding Jars
For use with MM 400 Mixer Mills
Marca: RETSCH 01.462.0215
Detalles adicionales : Weight : 0.43000kg
Descripción
Assorted sizes and materials
Especificaciones
Grinding Jar | |
35 mL | |
Ideal for leak-proof grinding | |
Zirconium Oxide | |
0.5 oz. |
1.18 oz. | |
Screw Top | |
MM 400 mixer mill | |
6 mm | |
15 mL |