Learn More
RETSCH ZM200 Ultra Centrifugal Mill: Ring Sieves
Stainless-steel sieves for use with ZM200 Ultra Centrifugal Mill
Brand: RETSCH 03.647.0245
Additional Details : Weight : 0.10000kg
Specifications
Ring Sieve with Reinforced Rim | |
Stainless Steel | |
Trapezoid holes |
ZM200 ultra centrifugal mil | |
0.12 mm |