Learn More
Ohaus™ Reusable Pan
OHAUS™ Series MB Reusable Pan is suitable for use with MB90 AM, MB120 AM, MB27 AM, and MB23 AM Moisture Analyzers.
Brand:  Ohaus™ 80252479
Additional Details : Weight : 1.00000kg
Product Code. 15535039
  • 116.00€ / 1 unit
Estimated Shipment: 31-05-2024
to see stock.
Description
Description
  • Accessory
  • For use with moisture analyzer
Specifications
Specifications
14 mm
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
3/Pk.
160 g