Learn More
Thermo Scientific™ Revco™ Plasma Freezers
Simplify operation and increase accuracy with these plasma freezers, designed to meet AABB, ANRC and FDA standards for safety and performance.
POA
Specifications
Type | Plasma Freezer |
---|---|
Temperature Range | -30°C |
Door Style | Solid |
Cabinet Material | High-impact, Powder-coated Paint |
Insulation | Urethane Foam |
Description
Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ Plasma Freezers. Our −30°C Plasma Freezers are designed to meet AABB, ANRC and FDA standards for safety and performance in plasma storage. Five models offer cabinet sizes to meet a variety of space needs and storage volume requirements.
Key Features- CFC-free refrigerant
- Preset temperature at -30°C
- Automatic condensate removal and defrost, with sensor that automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost build-up and optimizing coil efficiency
- Positive airflow systems
Microprocessor Control
- Icon-based controls and displays
- Preset temperature setpoint
- Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature and flashes bulb for undertemperature
- Large, easy-to-read digital display shows temperature within 1°C
- Audible/visual warnings for overtemperature, undertemperature or power failure
- Key-operated OFF/ON/ALARM ON master switch for setpoint security
- Push-button alarm test
- Remote alarm contacts
- Alarm silence, ringback and auto reset
- Controller battery backup system
Construction
- Cold-rolled steel cabinet surfaces finished with high-impact powder paint coating
- One solid door, hinged on right (except 51.1 cu. ft. model: two solid doors with keyed lock, self-close against closed-cell peripheral gasket)
- Full-length door handles
- Adjustable leveling feet
- High-density, CFC-free urethane foam insulation protects cabinet interior from ambient temperature fluctuations, minimizes operating costs
- Positive, forced-air circulation maintains uniformity at all shelf levels
- Directional airflow assures quick recovery after door openings
- Heavy-duty, hermetically sealed compressors
- 1 in. (2.5cm) access port with plug on undercounter model only
Ordering Information:
Alarm systems, temperature chart recorders and power monitors available separately. For additional voltages, contact your CSR.
Includes:
Space-saver and upright models come with a 6 in. (15.2cm) seven-day circular chart temperature recorder (optional free-standing on undercounter model) and extra-long probe for manual alarm testing.
Warranty varies by country, please contact us for detailsSpecifications
Plasma Freezer | |
Solid | |
Urethane Foam | |
AABB, ANRC, CE, cUL, FDA, TUV, UL | |
Warranty varies by country, please contact us for details | |
Alarm, Recorder | |
CFC-free |
-30°C | |
High-impact, Powder-coated Paint | |
Powder-coated Steel | |
Microprocessor | |
Automatic | |
Heavy-Duty, Industrial-Grade, Hermetically Sealed |