missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RG108

Product Code. p-7217469
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 17140042

Brand: STEMCELL Technologies 72212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Epigenetic modifier; Inhibits DNA methyltransferase (DNMT)

  • RG108 is an epigenetic modifier that inhibits DNA methyltransferase (IC50 = 115 nM)
  • RG108 is a non-nucleoside inhibitor that acts by direct binding to the methyltransferase enzyme whereby it blocks the enzyme active site
  • (Brueckner et al.), Stresemann et al.)
  • Enhances reprogramming efficiency of human and mouse somatic cells to induced pluripotent stem (iPS) cells (Mali et al.), Pasha et al.), Shi et al.)

Specifications

CAS 48208-26-0
Synonym N-Phthalyl-L-Tryptophan
Quantity 5 mg
Molecular Formula C19H14N2O4
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.