Learn More
IKA™ RH Digital Magnetic Stirrer
GREENER_CHOICE
Product Code. 13490547 Shop All IKA Products
Click to view available options
Capacity:
15 L
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 13490547
Brand: IKA™ 0005019800
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
An innovatively designed digital magnetic heated stirrer with bushing according to DIN 12878 for connecting an electronic contact thermometer, such as the ETS-D5.
  • Highly accurate temperature control is possible with sensor placed directly in the medium
  • A high heat output of 600 watts and stainless steel compound hot plate ensures rapid heating
  • Adjustable safety temperature from 100° to 360°C
  • Digital display of temperature and speed
  • Soft-start of stirring motor
  • Strong magnetic field and wide speed range for volumes up to 15 liters
  • Error code display
  • Easy operation with touch keypad
TRUSTED_SUSTAINABILITY
Specifications
Speed 100 to 2000 rpm
Stirring Positions 1
Top Plate Material Stainless Steel
Capacity 15 L
Display Digital LED
Voltage 100/115/230 V
Wattage 600 w
Height (Metric) 100 mm
Width (Metric) 160 mm
Frequency 50/60 Hz
Certifications/Compliance DIN EN 60529
Model RH Digital
Motor Type Electronically Controlled
Protection Class IP21
Speed Control Button
Temperature Range (Metric) 100°C to 360°C
Type RH Digital Magnetic Stirrer
Weight (Metric) 2.8 kg