Learn More
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings
Product Code. 11959214 Shop All Thermo Scientific Products
Click to view available options
Type:
Fitting Set, Short
Fitting Set, Standard Length
Fitting Set, X-Long
Replacement Ferrule
Unit Size:
Pack of 5
Product Code. 11959214
Brand: Thermo Scientific™ 6000066
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
  • Slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut
  • May be used on 1/16 in. metal or plastic tubing reliably up to 5000 psi
  • Reusable ferrule and nut
TRUSTED_SUSTAINABILITY
Specifications
Type Fitting Set, X-Long
Description 1/16 in. Extra-long fitting set; ea
For Use With (Equipment) PEEK Tubing
Material PEEK
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.