Learn More
Thermo Scientific™ Rheodyne™ 9725 and 9725i Sample Injectors
Product Code. 10762327 Shop All Thermo Scientific Products
Click to view available options
Description:
Continuous Flow
Continuous Flow, Position Sensing Switch
Unit Size:
Each
Product Code. 10762327
Brand: Thermo Scientific™ 9725
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ensure precise HPLC injections and protect against pressure shock with biocompatible dual-mode Thermo Scientific™ Rheodyne™ 9725 and 9725i Sample Injections.
Use biocompatible Thermo Scientific™ Rheodyne™ 7725 and 7725i Sample Injectors to maintain continuous HPLC flow between the load and inject positions and to protect against pressure shock. The Make-Before-Break (MBB) design ensures uninterrupted flow during sample injection. Constructed of Thermo Scientific™ PEEK™, these dual-mode injectors can use partial filling for zero sample waste or complete filling for better reproducibility. Inject from 1μL to 5mL samples with high accuracy and precision. The 9725i model features a position-sensing switch for a reproducible start signal. The injectors include a 20μL sample loop.
  • Biocompatible PEEK™ construction
  • Make-Before-Break (MBB) design
  • Can use partial filling for zero sample waste or complete filling for better reproducibility
  • Inject 1μL to 5mL with high accuracy and precision
  • 9725i features a position sensing switch for a reproducible start signal
Includes:20μL sample loop
TRUSTED_SUSTAINABILITY
Specifications
Mode Dual Mode
Volume (Metric) Sample 20 μL
Description Continuous Flow
For Use With (Equipment) HPLC
Type Continuous Flow HPLC Sample Injector
Stator Material PEEK
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.