Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Product Code. 10269514 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Rheodyne Injector 7030
Rheodyne Injector 8125
Rheodyne Injector 9010
Rheodyne Injector 9125
Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Rheodyne Injectors 7000/7010/7040
Rheodyne Injectors 7000/7010/7040/7067
Rheodyne Injectors 7060/7066
Rheodyne Injectors 7060/7066/9060
Rheodyne Injectors 7125/7126
Rheodyne Injectors 8125/8126
Material:
ETFE
PEEK
Vespel
missing translation for 'unitSize'
Each
Product Code. 10269514
missing translation for 'mfr': Thermo Scientific™ 7125079
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Rotor Seals
Material ETFE
Description 7125-079
For Use With (Equipment) Rheodyne Injectors 7125/7126
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.