Learn More
Thermo Scientific™ Rheodyne™ Stators
Product Code. 10353055 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Rheodyne Injector Models 7725
Rheodyne Injectors 7000/7010/7030/7040/7125
Rheodyne Injectors 7010-087/7125-081
Rheodyne Injectors 7060/7066
Rheodyne Injectors 7410/7413
Rheodyne Injectors 8125/8126
Rheodyne Injectors 9010/9030/9125
Unit Size:
Each
Product Code. 10353055
Brand: Thermo Scientific™ 9125043
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use these Thermo Scientific; Rheodyne; Stators for your Rheodyne Sample Injectors.
Use Thermo Scientific™ Rheodyne™ Stators with popular Rheodyne injector models used in high-performance liquid chromatography (HPLC) systems.
TRUSTED_SUSTAINABILITY
Specifications
Type Stators
Description For Rheodyne 9010/9030/9125
For Use With (Equipment) Rheodyne Injectors 9010/9030/9125
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.