Learn More
Rhodanine 99%, Thermo Scientific Chemicals
Catalog Number p-190976
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
141-84-4
C3H3NOS2
133.183
MFCD00005488
KIWUVOGUEXMXSV-UHFFFAOYSA-N
2-Thioxo-4-thiazolidinone
1201546
CHEBI:8830
2-sulfanylidene-1,3-thiazolidin-4-one
C1C(=O)NC(=S)S1
141-84-4
C3H3NOS2
133.183
MFCD00005488
KIWUVOGUEXMXSV-UHFFFAOYSA-N
2-Thioxo-4-thiazolidinone
1201546
CHEBI:8830
2-sulfanylidene-1,3-thiazolidin-4-one
C1C(=O)NC(=S)S1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 167°C to 170°C
Assay Percent Range 99%
Beilstein 27, 242
Merck Index 15, 8308
Infrared Spectrum Authentic
Formula Weight 133.2
Percent Purity ≥98.5%
Packaging Glass bottle
Color Beige to Yellow
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Rhodanine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Rhodanine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.