Learn More
Epredia™ Richard-Allan Scientific™ Eosin-Y Alcoholic, Eosin-Y with Phloxine, Eosin-Y Saturated, Eosin-Y Alcoholic
Shop All Epredia Products
Click to view available options
Quantity:
1 pt.
1 gal.
Type:
Alcoholic Eosin Y Stain
Eosin Phloxine Stain
Unit Size:
3.8 L
Pack of 4
Description
- Strict quality control of raw materials and finished product ensures a high quality product every time
- Eosin-Y Alcoholic produces three distinct color variations enhancing differentiation between muscle, red blood cells, and connective tissue
- Eosin-Y w/Phloxine yields a brighter reddish hue and a more dramatic differentiation of cytoplasmic color than Eosin-Y Alcoholic
- Eosin-Y Saturated is a very intense and rapid counterstain that can be used to make working solutions of Eosin-Y
Specifications
Specifications
| Type | Alcoholic Eosin Y Stain |
| Quantity | 1 pt. |
Safety and Handling
Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethyl alcohol, isopropyl,methyl alcohol,acetic acid, acid red 87