Learn More
Thermo Scientific Chemicals Rifapentine, 95%
Product Code. 15881052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15881052 25 mg 25mg
15801032 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 15881052 Supplier Thermo Scientific Chemicals Supplier No. 461900250
Request Bulk or Custom Format
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61379-65-5
Molecular Formula C47H64N4O12
Molecular Weight (g/mol) 877.03
InChI Key RISDZDVHYCURHD-QLWTZYDESA-N
PubChem CID 131846483
Specifications
Melting Point 179°C to 180°C
Assay Percent Range 94% min. (HPLC)
Quantity 25 mg
Formula Weight 877.03
Percent Purity 95%
Chemical Name or Material Rifapentine
compliance-icons
Product Identifier
  • Rifapentine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Rifapentine