Learn More
BINDER™ Access Port with Silicone Plug, Right Access
Product Code. 15873397 Shop All BINDER Products
Click to view available options
Material:
Silicone
Unit Size:
Each
Product Code. 15873397
Brand: BINDER™ 80121349
Only null left
Add to basket
Add to basket
BINDER™ Right Access Port is an accessory for use on the right of a variety of chambers and incubators.
  • For use on right
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 125 mm, right
For Use With (Equipment) BINDER Chambers Series MKF
Material Silicone
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 125 mm