Learn More
Quickfit™ Still Head Vertical Recovery Adapter
Product Code. 10438010
Change view
Click to view available options
Size:
19/26 (Cone)
24/29 (Condenser), 24/29 (Flask)
Unit Size:
Each
Pack of 5
Product Code. Size unitSize
10438010 24/29 (Condenser), 24/29 (Flask) Each
15507970 19/26 (Cone) Pack of 5
2 options
Product Code. 10438010
Brand: Quickfit SH2/33
Used in distillation assemblies in which the condenser is assembled in the vertical mode
Used in distillation assemblies in which the condenser is assembled in the vertical mode
  • Used in distillation assemblies in which the condenser is assembled in the vertical mode
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used in distillation assemblies in which the condenser is assembled in the vertical mode
Type Vertical Recovery Still Head
Material Borosilicate Glass
Certifications/Compliance ISO 3585
Size 24/29 (Condenser), 24/29 (Flask)