missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Ringers 085 R-Flex Impact Gloves

Product Code. p-7235728 Shop All Ansell Products
Click to view available options
Size:
10
11
12
3X-Large
7
8
9
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17318172

Brand: Ansell™ 08507

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These Ringers 085 R-Flex Impact Gloves provide innovative impact, cut, and cold protection combined with superior dexterity for ultimate comfort in a variety of end uses.

  • The Ringers 085 R-Flex Impact glove with Nitrile Half Dip and Fleece Lining combines protection against impact hazards and the highest levels of cut resistance, with added warmth for application in cold environments.
  • A breathable knit shell developed with Ringers' F3 Technology™ and patented TPR design for superior dexterity and comfort.
  • The sandy finish palm ensures secure grip on wet and dry surfaces, and touchscreen compatible index, middle and thumb tips allow technology interaction.
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material Nitrile
Color Multicolored
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material HPPE, Nitrile
Product Type Mechanical Protection Gloves
Size 7
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.