missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK5 Recombinant Human Protein
RIPK5 Recombinant Human Protein
Brand: Invitrogen PV4678
Additional Details : Weight : 0.00010kg
Description
RIPK5 is a serine⁄threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis.Specifications
NP_056190.1 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
RIPK5 Recombinant Human Protein | |
–80°C | |
1 Each | |
RIPK5 Recombinant Human Protein | |
RIPK | |
26nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 13.3μg/mL | |
GST Tag | |
Kinase and inhibitors | |
85% by SDS-PAGE |
0.46mg/mL | |
132.7kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
RIPK5, DustyPK | |
RIPK5 was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
RIPK5 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
For Research Use Only. Not for use in diagnostic procedures.