missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RNF213 Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
:
0.1mg
Description
The RNF213 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_665. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens RNF213, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction