Learn More
STEMCELL Technologies RoboSep™ Human NK Cell Isolation Kit
Product Code. 17177821
Encompass
Change view
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177821 1 x 109 Cells 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17177821 Supplier STEMCELL Technologies Supplier No. 17955RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ Human NK Cell Isolation Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This product can be used in place of the EasySep™ Human NK Cell Enrichment Kit (Catalog #19055) for even faster cell isolations
Specifications
Cell Type NK Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 Cells