missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROCK2 Recombinant Human Protein
ROCK2 is the predominant ROCK isoform in skeletal muscle.
1826.00€ - 7890.00€
Specifications
Accession Number | NP_004841 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 90.8kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408523
|
Invitrogen™
PR7116B |
100μg |
1826.00€
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10736413
|
Invitrogen™
PV4048 |
1mg |
7890.00€
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ROCK2 is an important regulator downstream of Rho GTPases.
Specifications
NP_004841 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
DMPK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
90.8kDa | |
RUO - research use only | |
ROCK2 | |
Human | |
GST Tag | |
Frozen |