Learn More
Rogo Sampaic™ Glass Coverslips
Product Code. 11983564
Click to view available options
Length (Metric):
18 mm
20 mm
22 mm
24 mm
Width (Metric):
18 mm
20 mm
22 mm
50 mm
60 mm
Quantity:
100/Pk.
1000/Pk.
200/Pk.
Unit Size:
Pack of 100
Pack of 1000
Pack of 200
Pack of 500
Product Code. 11983564
Brand: Rogo Sampaic™ BPD025
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Preserve and protect specimens on slides. Rogo Sampaic™ Glass Coverslips are manufactured from borosilicate glass for excellent transparency and prevention of color distortion.
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Product Type Cover Slide
Shape Rectangle
Material Borosilicate Glass
Length (Metric) 24 mm
Width (Metric) 60 mm
Dimensions (L x W) 24 x 60 mm
For Use With (Application) Slide microscopy
Certifications ISO 8255
Quantity 100/Pk.