Learn More
Eurofins DiscoverX ROKβ/ROCK-I Protein, Active
Description
Rho-associated, coiled-coil containing protein kinase (ROCK), including the ROCK1 and ROCK2 isoforms, is a protein kinase involved in signaling from Rho to actin cytoskeleton. PKA and the two ROCKs have been reported to have opposite effects on neurite formation. More studies are needed to provide significant insights into the role of ROCK in several pathological processes. ROCK inhibitors have been shown the benefit for treatment of various cancers, inflammation, coronary artery spasms and hypertension.
GenBank NM_005406
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 61.4 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥59% |
| Protein | ROCK-I, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.