missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ROR2 Recombinant Human Protein

ROR2 (receptor tyrosine kinase-like orphan receptor 2) is a type I transmembrane protein that belongs to the ROR subfamily of cell surface receptors.

1826.00€ - 7890.00€

Specifications

Accession Number NP_004551
For Use With (Application) Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Formulation 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Molecular Weight (g/mol) 85.2kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15478603
View Documents Promotion Details
Invitrogen™
PR7516A
100μg
1826.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 16-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10392213
View Documents Promotion Details
Invitrogen™
PV4179
1mg
7890.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 16-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ROR2 is thought to be involved in the early formation of the chondrocytes and cartilage and growth plate development.

Specifications

Specifications

NP_004551
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
ROR Kinase Family
Recombinant
Baculovirus
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
85.2kDa
RUO - research use only
ROR2
Human
GST Tag
Frozen
Videos
SDS
Documents

Documents

Product Certifications