Learn More
Abnova™ ROS1 Split FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | c-ros oncogene 1 , receptor tyrosine kinase |
| Gene Alias | MCF3,ROS,c-ros-1 |
| Gene Symbol | ROS1 |
| Gene ID (Entrez) | 6098 |
| Product Type | DNA/RNA |
| Quantity | 100 μL |
| Form | Liquid |