Learn More
Tocris Bioscience™ Rostafuroxin
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Rostafuroxin is a Na+/K+ ATPase modulator. Inhibits the Normalizes renal Na+/K+ ATPase pump function in mutant adducin- and endogenous ouabain-dependent forms of hypertension. Reduces blood pressure in Milan hypertensive rats. Controls mania-like behaviors in Myshkin mice having a Na+/K+ ATPase α3 subunit mutation. Antagonizes the effects of Ouabain (Cat.No. 1076) on myogenic tone and Ca2+ signaling in rat hippocampal neurons.
Specifications
Specifications
| Chemical Name or Material | 3β,5β,14β)-21,23-Epoxy-24-norchola-20,22-diene-3,14,17-triol |
| CAS | 156722-18-8 |
| Synonym | PST 2238 |
| Quantity | 10 mg |
| Target | Na+/K+ ATPase Modulators |
| Molecular Formula | C23H34O4 |
| Formula Weight | 374.51 |
| Purity | >98% |