Learn More
Fisherbrand™ Rotary Vane Vacuum Pump
Designed for laboratory applications requiring deep vacuum. Fisherbrand™ Rotary Vane Vacuum Pump is a two-stage direct driven rotary vane pumps that provide a high water vapor tolerance.
Brand:  Fisherbrand™
Additional Details : Weight : 17.55000kg
 View more versions of this product
Product Code. 12921141
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Two-stage direct driven rotary vane pump
  • Designed for wide range of physical and chemical laboratory applications and continuous running
  • Low weight, small footprint and high pumping speed, increased chemical resistance are advantageous features in research and routine processes
  • Rugged reliable design, combined with low noise level and ease of use
  • Innovative gas ballast valve prevents moisture condensing and high tolerance of water and solvent vapors
  • Intake line is vacuum sealed by and anti suckback valve to protect your application from undesired venting and oil back flow after pump is turned off
  • Vertical oil sight glass allows easy monitoring of oil level and condition
Specifications
Specifications
2 x 10-3 mbar
43 x 15 x 23.5 cm (16.9 x 5.9 x 9.25 in.)
42.9 lb.
97/110 L/min.
50/60 Hz
97 L/min.
230 V
115/230 V 50/60 Hz motor with Schuko UK and US power leads
90 w
19.5 kg
Labovac 10
Rotary Vane Vacuum Pump
2 x 10-3 mbar