Learn More
OHAUS™ Rotor, Angle, 12 x 15ml RB/FA, ID
Shop All Ohaus Products
Click to view available options
Capacity:
12 x 15 mL
Unit Size:
Each
Description
Requires
Frontier 5000 Series Multi Pro Centrifuge
- Capacity: 12 x 15 ml
- Max g-force: 4,427g
- Max speed: 6,000 rpm
Warranty
2 years
Specifications
Specifications
| Description | Rotor, Angle, 12x15ml RB/FA, ID |
| Material | Polypropylene |
| Type | Angle Rotor |
| Capacity | 12 x 15 mL |
| Max. RCF | 4,427 x g |
| Max. Speed | 6,000 rpm |
| Net Weight | 2.740 kg |
| For Use With (Equipment) | FC5714 |
| Angle | 32° |
| Includes | NA |
| Show More |