Learn More
Eppendorf™ Rotor F-48x15, Plus 48 Steel Sleeves
Shop All Eppendorf Products
Click to view available options
Capacity:
40 x 15 mL Conical and Glass Tubes
Unit Size:
Each
Description
- Suitable for use with 15 mL conical and glass tubes
- Includes steel sleeves and adapters
- Suitable for use with 5910 R Centrifuge
Specifications
Specifications
| Capacity | 40 x 15 mL Conical and Glass Tubes |
| Max. Speed | 5,500 rpm |
| For Use With (Equipment) | 5910R Centrifuge |
| Includes | 48 steel sleeves and adapters |
| No. of Places | 40 |