Learn More
Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R
Product Code. 15768369 Shop All Eppendorf Products
Click to view available options
Capacity:
20 × 5 mL
4 to 750 mL
4 x 1,000 mL
4 x 750 mL
48 x 2 mL
6 x 250 mL
6 × 50 mL
Max. Speed:
10,100 rpm
13,100 rpm
13,500 rpm
13,700 rpm
4,500 rpm
4,700 rpm
4500 rpm
Max. RCF:
15,054 x g
20,130 x g
20,913 x g
21,194 x g
4,347 x g
4,800 x g
Includes:
Eppendorf QuickLock™ Rotor Lid
Rotor Lid
Round Buckets
Universal Buckets
Without Buckets
Unit Size:
Each
Product Code. 15768369
Brand: Eppendorf™ 5895121004
Eppendorf Rotor for Centrifuge 5910 Ri and 5920 R-precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds, buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor S-4x750, without buckets
Capacity 4 x 750 mL
Max. RCF 4,800 x g
Max. Speed 4,700 rpm
For Use With (Equipment) Centrifuges 5910 Ri and 5920 R
Includes Without Buckets
Model S-4x750, Aerosol-tight