Learn More
IKA™ Round Bottom Flask Inlay
Shop All IKA Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
500 mL
Unit Size:
Each
Description
- Round bottom
- Anodized aluminum construction
- Temperature resistant
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Shape | Round Bottom |
| Temperature (Metric) | 180°C |
| Type | Flask Carrier |