Learn More
IKA™ Round Bottom Flask Inlay
Product Code. 15213305 Shop All IKA Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
500 mL
Unit Size:
Each
Product Code. 15213305
Brand: IKA™ 0025000636
Only null left
Add to basket
Add to basket
Round bottom flask inlay constructed of anodized aluminum
  • Round bottom
  • Anodized aluminum construction
  • Temperature resistant
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 10 mL
Diameter (English) 1.3 in.
Diameter (Metric) 34 mm
Shape Round Bottom
Temperature (Metric) 180°C
Type Flask Inlay